missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chlorobenzenesulfonamide, 98%, Thermo Scientific Chemicals
£35.20 - £126.00
Chemical Identifiers
CAS | 17260-71-8 |
---|---|
Molecular Formula | C6H6ClNO2S |
Molecular Weight (g/mol) | 191.63 |
MDL Number | MFCD00051976 |
InChI Key | WSYQJNPRQUFCGL-UHFFFAOYSA-N |
Synonym | benzenesulfonamide, 3-chloro, 3-chlorobenzene-1-sulfonamide, 3-chlorobenzenesulphonamide, 3-chloro-benzenesulfonamide, m-chlorobenzenesulfonamide, acmc-20a3ig, 3-chlorobenzensulfonamide, 3-chloranylbenzenesulfonamide, benzenesulfonamide,3-chloro, benzenesulfonamide, 3-chloro-9ci |
PubChem CID | 519377 |
IUPAC Name | 3-chlorobenzenesulfonamide |
SMILES | NS(=O)(=O)C1=CC=CC(Cl)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11348105
|
Thermo Scientific Alfa Aesar
L08349.03 |
1 g |
£35.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11358105
|
Thermo Scientific Alfa Aesar
L08349.06 |
5 g |
£126.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It can react with 1-chloro-4-isocyanato-benzene to prepare C13H10Cl2N2O3S in the presence of 1 N aq. NaOH and solvent acetone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt can react with 1-chloro-4-isocyanato-benzene to prepare C13H10Cl2N2O3S in the presence of 1 N aq. NaOH and solvent acetone.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
17260-71-8 | |
191.63 | |
WSYQJNPRQUFCGL-UHFFFAOYSA-N | |
519377 | |
NS(=O)(=O)C1=CC=CC(Cl)=C1 |
C6H6ClNO2S | |
MFCD00051976 | |
benzenesulfonamide, 3-chloro, 3-chlorobenzene-1-sulfonamide, 3-chlorobenzenesulphonamide, 3-chloro-benzenesulfonamide, m-chlorobenzenesulfonamide, acmc-20a3ig, 3-chlorobenzensulfonamide, 3-chloranylbenzenesulfonamide, benzenesulfonamide,3-chloro, benzenesulfonamide, 3-chloro-9ci | |
3-chlorobenzenesulfonamide |
Specifications
17260-71-8 | |
C6H6ClNO2S | |
2832245 | |
Slightly soluble in water. | |
NS(=O)(=O)C1=CC=CC(Cl)=C1 | |
191.63 | |
191.64 | |
3-Chlorobenzenesulfonamide |
144°C to 148°C | |
MFCD00051976 | |
benzenesulfonamide, 3-chloro, 3-chlorobenzene-1-sulfonamide, 3-chlorobenzenesulphonamide, 3-chloro-benzenesulfonamide, m-chlorobenzenesulfonamide, acmc-20a3ig, 3-chlorobenzensulfonamide, 3-chloranylbenzenesulfonamide, benzenesulfonamide,3-chloro, benzenesulfonamide, 3-chloro-9ci | |
WSYQJNPRQUFCGL-UHFFFAOYSA-N | |
3-chlorobenzenesulfonamide | |
519377 | |
98% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only