missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11444013
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2888-06-4
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.06
MDL Number MFCD00051697
InChI Key OINWZUJVEXUHCC-UHFFFAOYSA-N

Specifications

Melting Point ∼21°C
Density 1.499
Boiling Point 102°C to 104°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.569
Quantity 1 g
UN Number UN3265
Beilstein 2937139
Sensitivity Moisture sensitive
Synonym 3-chlorobenzene-1-sulfonyl chloride, 3-chlorobenzenesulphonyl chloride, 3-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 3-chloro, m-chlorobenzenesulfonyl chloride, 3-chloro-benzenesulfonyl chloride, 3-chlorophenylsulfonyl chloride, pubchem5076, m-chlorbenzolsulfochlorid, acmc-1cj4b
SMILES C1=CC(=CC(=C1)Cl)S(=O)(=O)Cl
IUPAC Name 3-chlorobenzenesulfonyl chloride
PubChem CID 17909
Formula Weight 211.07
Percent Purity 98%
Chemical Name or Material 3-Chlorobenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'