Learn More
3-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals
£14.10 - £243.00
Chemical Identifiers
CAS | 618-46-2 |
---|---|
Molecular Formula | C7H4Cl2O |
Molecular Weight (g/mol) | 175.01 |
MDL Number | MFCD00000671 |
InChI Key | WHIHIKVIWVIIER-UHFFFAOYSA-N |
Synonym | m-chlorobenzoyl chloride, benzoyl chloride, 3-chloro, benzoyl chloride, m-chloro, m-chlorobenzoylchloride, 3-chlorobenzoylchloride, benzoylchloride, 3-chloro, mcoc, 3-chlorobenzoic acid chloride, ccris 8615, 3chlorobenzoyl chloride |
PubChem CID | 69252 |
IUPAC Name | 3-chlorobenzoyl chloride |
SMILES | ClC(=O)C1=CC=CC(Cl)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10172610
|
Thermo Scientific Acros
108740050 |
5 mL | Glass bottle |
£14.10
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10010840
|
Thermo Scientific Acros
108740500 |
50 mL | Glass bottle |
£39.80
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10020840
|
Thermo Scientific Acros
108741000 |
100 mL | Glass bottle |
£63.90
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10030850
|
Thermo Scientific Acros
108745000 |
500 mL | Glass bottle |
£243.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
618-46-2 | |
175.01 | |
WHIHIKVIWVIIER-UHFFFAOYSA-N | |
69252 | |
ClC(=O)C1=CC=CC(Cl)=C1 |
C7H4Cl2O | |
MFCD00000671 | |
m-chlorobenzoyl chloride, benzoyl chloride, 3-chloro, benzoyl chloride, m-chloro, m-chlorobenzoylchloride, 3-chlorobenzoylchloride, benzoylchloride, 3-chloro, mcoc, 3-chlorobenzoic acid chloride, ccris 8615, 3chlorobenzoyl chloride | |
3-chlorobenzoyl chloride |
Especificaciones
618-46-2 | |
100.0 | |
1.3600g/mL | |
110°C | |
0.03% max. Total other poly-halogenated acid halides (GC) | |
ClC6H4COCl | |
MFCD00000671 | |
1.36 | |
Solubility in water: decomposes. Other solubilities: soluble in alcohol,ether and acetone | |
ClC(=O)C1=CC=CC(Cl)=C1 | |
175.01 | |
175.01 | |
Liquid |
99.0 | |
Colorless | |
223.0°C to 225.0°C | |
Authentic | |
C7H4Cl2O | |
1.5680 to 1.5700 | |
09, 338 | |
m-chlorobenzoyl chloride, benzoyl chloride, 3-chloro, benzoyl chloride, m-chloro, m-chlorobenzoylchloride, 3-chlorobenzoylchloride, benzoylchloride, 3-chloro, mcoc, 3-chlorobenzoic acid chloride, ccris 8615, 3chlorobenzoyl chloride | |
WHIHIKVIWVIIER-UHFFFAOYSA-N | |
3-chlorobenzoyl chloride | |
69252 | |
99+% | |
3-Chlorobenzoyl chloride, 99+% |
Seguridad y manipulación
GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 210-552-6
missing translation for 'tsca' : TSCA
- 3-Chlorobenzoyl chloride
Palabra de advertencia
- Danger
Clasificación
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Indicaciones de peligro
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- EUH029-Contact with water liberates toxic gas.
Consejos de prudencia
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only