missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzyl bromide, 97%, Thermo Scientific Chemicals

Product Code. 11492528
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3-Chlorobenzyl bromide was used in the synthesis of symmetrical and unsymmetrical benzyl thioethers. It was used as starting reagent during the synthesis of 1-(3-chlorobenzyl)-2-(pyrrolidin-1-ylmethyl)-1H-benzimidazole dihydrochloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chlorobenzyl bromide was used in the synthesis of symmetrical and unsymmetrical benzyl thioethers. It was used as starting reagent during the synthesis of 1-(3-chlorobenzyl)-2-(pyrrolidin-1-ylmethyl)-1H-benzimidazole dihydrochloride.

Solubility
Decomposes in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-80-3
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.48
MDL Number MFCD00000597
InChI Key LZIYAIRGDHSVED-UHFFFAOYSA-N
Synonym 3-chlorobenzyl bromide, 1-bromomethyl-3-chlorobenzene, 3-chlorobenzylbromide, alpha-bromo-3-chlorotoluene, m-chlorobenzyl bromide, benzene, 1-bromomethyl-3-chloro, alpha-bromo-m-chlorotoluene, .alpha.-bromo-m-chlorotoluene, 1-bromomethyl-3-chloro-benzene, 3-chorobenzylbromide
PubChem CID 69838
IUPAC Name 1-(bromomethyl)-3-chlorobenzene
SMILES ClC1=CC=CC(CBr)=C1

Specifications

Density 1.567
Boiling Point 108°C to 110°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.589
Quantity 10 g
UN Number UN3265
Beilstein 636504
Solubility Information Decomposes in water.
Formula Weight 205.49
Percent Purity 97%
Chemical Name or Material 3-Chlorobenzyl bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'