missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloroperoxybenzoic acid, 50-55%, cont. ca 10% 3-chlorobenzoic acid, balance water, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
3-Chloroperoxybenzoic acid is a strong oxidizing agent used in the oxidation reactions such as aldehydes and ketones to esters (Bayer-Villiger-Oxidation), olefines to epoxides, sulfides to sulfoxides and sulfones, and amines to nitroalkanes, N-oxides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Chloroperoxybenzoic acid is a strong oxidizing agent used in the oxidation reactions such as aldehydes and ketones to esters (Bayer-Villiger-Oxidation), olefines to epoxides, sulfides to sulfoxides and sulfones, and amines to nitroalkanes, N-oxides.
Solubility
Soluble in potassium hydroxide, chloroform and acetone. Insoluble in water.
Notes
Heat sensitive. Incompatible with reactive with reducing agents, combustible materials, organic materials and alkalies.
3-Chloroperoxybenzoic acid is a strong oxidizing agent used in the oxidation reactions such as aldehydes and ketones to esters (Bayer-Villiger-Oxidation), olefines to epoxides, sulfides to sulfoxides and sulfones, and amines to nitroalkanes, N-oxides.
Solubility
Soluble in potassium hydroxide, chloroform and acetone. Insoluble in water.
Notes
Heat sensitive. Incompatible with reactive with reducing agents, combustible materials, organic materials and alkalies.
Specifications
Melting Point | 64°C to 66°C |
Quantity | 25 g |
UN Number | UN3106 |
Beilstein | 608317 |
Solubility Information | Soluble in potassium hydroxide,chloroform and acetone. Insoluble in water. |
Formula Weight | 172.57 |
Percent Purity | 50 to 55% |
Assay | cont. ≈10% 3-chlorobenzoic acid, balance water |
Chemical Name or Material | 3-Chloroperoxybenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'