Learn More
3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
3-Chloroperoxybenzoic acid, 70-75%, C7H5ClO3, CAS Number-937-14-4, 535-80-8, 7732-18-5
Brand: Thermo Scientific Acros 255795000
167.25 GBP valid until 2024-07-31
Use promo code "23251" to get your promotional price.
Code : 29
Additional Details : CAS Number : 937-14-4 Weight : 0.50000kg Transport : UN number : 3106 Chem class : 5.2
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
3-Chloroperoxybenzoic Acid | |
937-14-4,535-80-8,7732-18-5 | |
30.0 | |
White | |
di-m-Chlorobenzoyl peroxide: < 0.1% Typical (HPLC) | |
C7H5ClO3 | |
MFCD00002127 | |
3106 | |
01,135; 02,68; 03,49; 04,85; 05,120; 06,110; 07,62; 08,97; 09,108; 10,92; 11,122; 12,118; 13,76; 15,86; 16,80; 17,76 | |
15,2154 | |
Solubility in water: insoluble. Other solubilities: soluble in koh, soluble in chloroform and acetone | |
OOC(=O)C1=CC=CC(Cl)=C1 | |
172.56 | |
CHEBI:52091 | |
70-75% |
70 - 75% | |
25.0 | |
92.0°C to 94.0°C | |
0.5600g/mL | |
Plastic bottle | |
ClC6H4CO3H | |
500 g | |
09,IV,972 | |
0.56 | |
3-chloroperoxybenzoic acid, 3-chloroperbenzoic acid, mcpba, m-chloroperbenzoic acid, m-chloroperoxybenzoic acid, meta-chloroperoxybenzoic acid, benzenecarboperoxoic acid, 3-chloro, m-chlorobenzoyl hydroperoxide, meta-chloroperbenzoic acid, 3-chlorobenzoperoxoic acid | |
NHQDETIJWKXCTC-UHFFFAOYSA-N | |
3-chlorobenzenecarboperoxoic acid | |
70297 | |
172.57 | |
Moist Powder |
Safety and Handling
- 3-Chloroperoxybenzoic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Organic peroxide Type C & D
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H242-Heating may cause a fire.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only