Learn More
3-Chlorophenol, 98+%, Thermo Scientific Chemicals
£54.40 - £467.00
Chemical Identifiers
CAS | 108-43-0 |
---|---|
Molecular Formula | C6H5ClO |
Molecular Weight (g/mol) | 128.56 |
MDL Number | MFCD00002256 |
InChI Key | HORNXRXVQWOLPJ-UHFFFAOYSA-N |
Synonym | m-chlorophenol, phenol, 3-chloro, phenol, m-chloro, 3-hydroxychlorobenzene, m-chlorophenic acid, meta-chlorophenol, unii-z2z7m2ftad, 3-chloro-1-hydroxybenzene, 3-chloro-phenol, ccris 641 |
PubChem CID | 7933 |
ChEBI | CHEBI:38855 |
IUPAC Name | 3-chlorophenol |
SMILES | OC1=CC=CC(Cl)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11449050
|
Thermo Scientific Alfa Aesar
A13671.18 |
50 g |
£54.40
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11469050
|
Thermo Scientific Alfa Aesar
A13671.22 |
100 g |
£93.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459050
|
Thermo Scientific Alfa Aesar
A13671.30 |
250 g |
£188.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439050
|
Thermo Scientific Alfa Aesar
A13671.0B |
1000 g | |||||||
Description
3-Chlorophenol is widely used in the pharmaceutical, dyes and organic synthesis industry. It is an important intermediate in the synthesis of chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Chlorophenol is widely used in the pharmaceutical, dyes and organic synthesis industry. It is also used as a disinfectant. It is an important intermediate in the synthesis of chemicals.
Solubility
Soluble in ethanol and ether.
Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
108-43-0 | |
128.56 | |
HORNXRXVQWOLPJ-UHFFFAOYSA-N | |
7933 | |
3-chlorophenol |
C6H5ClO | |
MFCD00002256 | |
m-chlorophenol, phenol, 3-chloro, phenol, m-chloro, 3-hydroxychlorobenzene, m-chlorophenic acid, meta-chlorophenol, unii-z2z7m2ftad, 3-chloro-1-hydroxybenzene, 3-chloro-phenol, ccris 641 | |
CHEBI:38855 | |
OC1=CC=CC(Cl)=C1 |
Specifications
108-43-0 | |
1.245 | |
120°C (248°F) | |
1.566 | |
UN2020 | |
14,2154 | |
Soluble in ethanol and ether. | |
OC1=CC=CC(Cl)=C1 | |
128.56 | |
CHEBI:38855 | |
≥98% |
30°C to 34°C | |
214°C | |
C6H5ClO | |
MFCD00002256 | |
1634401 | |
m-chlorophenol, phenol, 3-chloro, phenol, m-chloro, 3-hydroxychlorobenzene, m-chlorophenic acid, meta-chlorophenol, unii-z2z7m2ftad, 3-chloro-1-hydroxybenzene, 3-chloro-phenol, ccris 641 | |
HORNXRXVQWOLPJ-UHFFFAOYSA-N | |
3-chlorophenol | |
7933 | |
128.56 | |
3-Chlorophenol |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID
missing translation for 'einecsNumber' : 203-582-6
missing translation for 'rtecsNumber' : SK2450000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only