missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropionyl chloride, 98%

Product Code. 10520181 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10520181

Brand: Thermo Scientific Acros 109991000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 625-36-5 | C3H4Cl2O | 126.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-36-5
Molecular Formula C3H4Cl2O
Molecular Weight (g/mol) 126.96
MDL Number MFCD00000747
InChI Key INUNLMUAPJVRME-UHFFFAOYSA-N
Synonym 3-chloropropionyl chloride, propanoyl chloride, 3-chloro, propionyl chloride, 3-chloro, 3-chloropropionylchloride, beta-chloropropionyl chloride, 3-chloropropionic acid chloride, chloropropionyl chloride, propanoylchloride, 3-chloro, .beta.-chloropropanoyl chloride, .beta.-chloropropionyl chloride
PubChem CID 69364
IUPAC Name 3-chloropropanoyl chloride
SMILES ClCCC(Cl)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -32.0°C
Color Brown to Colorless
Density 1.3300g/mL
Boiling Point 143.0°C to 145.0°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CH2COCl
Refractive Index 1.4560 to 1.4580
Quantity 100 mL
Beilstein 02, 250
Specific Gravity 1.33
Solubility Information Solubility in water: reacts. Other solubilities: reacts violently with alcohol,amines,other,nucleophtilic solvents
Viscosity 1.4 mPa.s (20°C)
Formula Weight 126.97
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Chloropropionyl chloride, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Chloropropionyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.