missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropionyl chloride, 98%, Thermo Scientific Chemicals

£38.09 - £186.93

Chemical Identifiers

CAS 625-36-5
Molecular Formula C3H4Cl2O
Molecular Weight (g/mol) 126.96
MDL Number MFCD00000747
InChI Key INUNLMUAPJVRME-UHFFFAOYSA-N
Synonym 3-chloropropionyl chloride, propanoyl chloride, 3-chloro, propionyl chloride, 3-chloro, 3-chloropropionylchloride, beta-chloropropionyl chloride, 3-chloropropionic acid chloride, chloropropionyl chloride, propanoylchloride, 3-chloro, .beta.-chloropropanoyl chloride, .beta.-chloropropionyl chloride
PubChem CID 69364
IUPAC Name 3-chloropropanoyl chloride
SMILES ClCCC(Cl)=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10520181
View Documents
Thermo Scientific Acros
109991000
100 mL Glass bottle
£38.09
100mL
Please to purchase this item. Need a web account? Register with us today!
10408830
View Documents Promotion Details
Thermo Scientific Acros
109995000
500 mL Glass bottle
£186.93
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

625-36-5
126.96
INUNLMUAPJVRME-UHFFFAOYSA-N
69364
ClCCC(Cl)=O
C3H4Cl2O
MFCD00000747
3-chloropropionyl chloride, propanoyl chloride, 3-chloro, propionyl chloride, 3-chloro, 3-chloropropionylchloride, beta-chloropropionyl chloride, 3-chloropropionic acid chloride, chloropropionyl chloride, propanoylchloride, 3-chloro, .beta.-chloropropanoyl chloride, .beta.-chloropropionyl chloride
3-chloropropanoyl chloride

Specifications

625-36-5
100.0
Brown to Colorless
143.0°C to 145.0°C
Authentic
C3H4Cl2O
1.4560 to 1.4580
02, 250
3-chloropropionyl chloride, propanoyl chloride, 3-chloro, propionyl chloride, 3-chloro, 3-chloropropionylchloride, beta-chloropropionyl chloride, 3-chloropropionic acid chloride, chloropropionyl chloride, propanoylchloride, 3-chloro, .beta.-chloropropanoyl chloride, .beta.-chloropropionyl chloride
INUNLMUAPJVRME-UHFFFAOYSA-N
3-chloropropanoyl chloride
69364
126.97
Liquid
97.5
-32.0°C
1.3300g/mL
49°C
97.5% min. (GC)
ClCH2CH2COCl
MFCD00000747
1.33
Solubility in water: reacts. Other solubilities: reacts violently with alcohol,amines,other,nucleophtilic solvents
ClCCC(Cl)=O
126.96
1.4 mPa.s (20°C)
98%
3-Chloropropionyl chloride, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.
May be corrosive to metals.
Corrosive to the respiratory tract.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a POISON CENTER or doctor/physi

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 210-890-4

missing translation for 'tsca' : TSCA

Product Identifier
  • 3-Chloropropionyl chloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
  • EUH071-Corrosive to the respiratory tract.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only