missing translation for 'onlineSavingsMsg'
Learn More

(3-Chloropropyl)trimethoxysilane, 97+%, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 11354568
Click to view available options
Quantity:
50 mL
250 mL
1 L
Unit Size:
1 L
250mL
50mL
This item is not returnable. View return policy

Product Code. 11354568

Brand: Thermo Scientific Alfa Aesar 042413.AD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2530-87-2 | C6H15ClO3Si | 198.718 g/mol

(3-Chloropropyl)trimethoxysilane is used as adhesives and sealant chemicals. It is also used in the preparation of 3-iodopropyltrimethoxysilane by reacting with sodium iodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2530-87-2
Molecular Formula C6H15ClO3Si
Molecular Weight (g/mol) 198.718
MDL Number MFCD00000997
InChI Key OXYZDRAJMHGSMW-UHFFFAOYSA-N
Synonym 3-chloropropyl trimethoxysilane, silane, 3-chloropropyl trimethoxy, 3-chloropropyltrimethyoxysilane, sila-ace s 620, cps-m, silane 3-chloropropyl tris methoxy, 3-trimethoxysilyl propyl chloride, unii-t21bnl1s7f, cptmo, 3-chloropropyl trimethoxysilan
PubChem CID 62449
IUPAC Name 3-chloropropyl(trimethoxy)silane
SMILES CO[Si](CCCCl)(OC)OC

Specifications

Boiling Point 195°C to 197°C
Melting Point -50°C
Physical Form Liquid
Quantity 50 mL
Beilstein 969627
Flash Point 78°C (172°F)
Solubility Information Miscible with organic solvents.
Formula Weight 198.72
Percent Purity ≥97%
Odor Recognizable
Packaging ChemSeal™ bottles
Refractive Index 1.42
Sensitivity Moisture sensitive
Density 1.077 g/mL
Chemical Name or Material (3-Chloropropyl)trimethoxysilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.