missing translation for 'onlineSavingsMsg'
Learn More

(3-Chloropropyl)trimethylsilane, 98%, Thermo Scientific Chemicals

£36.70 - £122.00

Chemical Identifiers

CAS 2344-83-4
Molecular Formula C6H15ClSi
Molecular Weight (g/mol) 150.72
MDL Number MFCD00045299
InChI Key QXDDDCNYAAJLBT-UHFFFAOYSA-N
Synonym 3-chloropropyl trimethylsilane, silane, 3-chloropropyl trimethyl, 3-chloropropyl trimethyl silane, acmc-20apo8, chloropropyltrimethylsilane, chloropropyl trimethyl silane, 3 3-chloropropyl trimethylsilane, gamma-chloropropyl trimethylsilane
PubChem CID 75362
IUPAC Name 3-chloropropyl(trimethyl)silane
SMILES C[Si](C)(C)CCCCl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11371587
View Documents
Thermo Scientific Alfa Aesar
L04458.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11381587
View Documents
Thermo Scientific Alfa Aesar
L04458.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

(3-Chloropropyl)trimethylsilane used in the preparation of: 8-(3-trimethylsilylpropoxy)quinolone under phase transfer catalytic conditions, 5-trimethylsilyl pentanol, N-(tri-methylsilylalkyl)diamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Chloropropyl)trimethylsilane used in the preparation of: 8-(3-trimethylsilylpropoxy)quinolone under phase transfer catalytic conditions, 5-trimethylsilyl pentanol, N-(tri-methylsilylalkyl)diamines.

Solubility
Not miscible in water.

Notes
Store under inert gas. Moisture sensitive. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

2344-83-4
150.72
QXDDDCNYAAJLBT-UHFFFAOYSA-N
75362
C[Si](C)(C)CCCCl
C6H15ClSi
MFCD00045299
3-chloropropyl trimethylsilane, silane, 3-chloropropyl trimethyl, 3-chloropropyl trimethyl silane, acmc-20apo8, chloropropyltrimethylsilane, chloropropyl trimethyl silane, 3 3-chloropropyl trimethylsilane, gamma-chloropropyl trimethylsilane
3-chloropropyl(trimethyl)silane

Specifications

150°C to 152°C
C6H15ClSi
UN1993
49°C (120°F)
Not miscible in water.
C[Si](C)(C)CCCCl
150.72
150.73
1.432
(3-Chloropropyl)trimethylsilane
2344-83-4
MFCD00045299
1732625
3-chloropropyl trimethylsilane, silane, 3-chloropropyl trimethyl, 3-chloropropyl trimethyl silane, acmc-20apo8, chloropropyltrimethylsilane, chloropropyl trimethyl silane, 3 3-chloropropyl trimethylsilane, gamma-chloropropyl trimethylsilane
QXDDDCNYAAJLBT-UHFFFAOYSA-N
3-chloropropyl(trimethyl)silane
75362
98%
0.879
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'einecsNumber' : 219-059-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only