missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropylamine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11448396
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6276-54-6
Molecular Formula C3H9Cl2N
Molecular Weight (g/mol) 130.012
MDL Number MFCD00012913
InChI Key IHPRVZKJZGXTBQ-UHFFFAOYSA-N
Synonym 3-chloropropylamine hydrochloride, 3-chloropropan-1-amine hydrochloride, 3-chloropropylamine hcl, 1-amino-3-chloropropane hydrochloride, 3-chloro-1-propanamine hydrochloride, 1-propanamine, 3-chloro-, hydrochloride, 3-chloropropylammonium chloride, acmc-1b7ws, chloropropylamine hydrochloride, ksc491e7b
PubChem CID 11469095
IUPAC Name 3-chloropropan-1-amine;hydrochloride
SMILES C(CN)CCl.Cl

Specifications

Melting Point 147°C to 152°C
Quantity 1000 g
Beilstein 3667451
Formula Weight 130.02
Percent Purity 98%
Chemical Name or Material 3-Chloropropylamine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'