missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorotoluene, 97%, Thermo Scientific Chemicals

Product Code. 10468340
Click to view available options
Quantity:
100 mL
250 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-41-8
Molecular Formula C7H7Cl
Molecular Weight (g/mol) 126.58
MDL Number MFCD00000595
InChI Key OSOUNOBYRMOXQQ-UHFFFAOYSA-N
Synonym 3-chlorotoluene, m-chlorotoluene, benzene, 1-chloro-3-methyl, m-tolyl chloride, 3-chloro-1-methylbenzene, 1-methyl-3-chlorobenzene, toluene, m-chloro, meta-chlorotoluene, unii-5k8172k9tt, chlorotoluene 3-
PubChem CID 7931
IUPAC Name 1-chloro-3-methylbenzene
SMILES CC1=CC=CC(Cl)=C1

Specifications

Melting Point -48°C
CAS Min % 96.0
Color Colorless to Yellow
CAS Max % 100.0
Density 1.0700g/mL
Boiling Point 160°C to 162°C
Quantity 250 mL
Flash Point 50°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5210 to 1.523
Assay Percent Range 97%
Specific Gravity 1.07
Linear Formula CH3C6H4Cl
Beilstein 05, 291
Merck Index 15, 2175
Solubility Information Solubility in water: slightly soluble
Formula Weight 126.59
Percent Purity 97%
Chemical Name or Material 3-Chlorotoluene, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Chlorotoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 3
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'