missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-4-fluorobenzenesulfonyl chloride, 98%, Thermo Scientific™

Product Code. 11319905
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351003-23-1
Molecular Formula C7H3ClFNO2S
Molecular Weight (g/mol) 219.614
MDL Number MFCD03093760
InChI Key KDVZADPGGPBAAK-UHFFFAOYSA-N
Synonym 4-fluoro-3-cyanobenzenesulfonyl chloride, 3-cyano-4-fluorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 3-cyano-4-fluoro, 3-cyano-4-fluorobenzenesulphonyl chloride, 3-cyano-4-fluoro-benzenesulfonyl chloride, 4-fluoro-3-cyanobenzenesulfonylchloride, 4-fluoro-3-cyanobenzenesulphonyl chloride, 5-chlorosulfonyl-2-fluorobenzenecarbonitrile, pubchem5110, acmc-20a3ve
PubChem CID 2778908
IUPAC Name 3-cyano-4-fluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)C#N)F

Specifications

Melting Point 67°C to 70°C
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 219.62
Percent Purity 98%
Chemical Name or Material 3-Cyano-4-fluorobenzenesulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.