missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyano-4-methoxy-2-pyridone, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Used as a intermediate for gimeracil and pharmaceutical application.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as a intermediate for gimeracil and pharmaceutical application.
Solubility
Soluble in Dimethyl sulfoxide.
Notes
Hydroscopic. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from water/moisture and oxidizing agent.
Used as a intermediate for gimeracil and pharmaceutical application.
Solubility
Soluble in Dimethyl sulfoxide.
Notes
Hydroscopic. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from water/moisture and oxidizing agent.
Specifications
Specifications
Melting Point | 278°C |
Color | Yellow |
Quantity | 25 g |
UN Number | UN3439 |
Beilstein | 5929290 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in Dimethyl sulfoxide. |
IUPAC Name | 4-methoxy-2-oxo-1H-pyridine-3-carbonitrile |
Formula Weight | 150.14 |
Percent Purity | 95% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'