missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzenesulfonamide, 98%, Thermo Scientific™

Product Code. 11466610
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3118-68-1
Molecular Formula C7H6N2O2S
Molecular Weight (g/mol) 182.197
MDL Number MFCD02089633
InChI Key LTOFQPCBIZPXFH-UHFFFAOYSA-N
PubChem CID 2808718
IUPAC Name 3-cyanobenzenesulfonamide
SMILES C1=CC(=CC(=C1)S(=O)(=O)N)C#N

Specifications

Melting Point 151°C to 153°C
Quantity 1 g
UN Number UN3439
Synonym 3-cyanobenzene-1-sulfonamide, benzenesulfonamide, 3-cyano, benzenesulfonamide, 3-cyano-9ci, 3-sulfamoylbenzonitrile, acmc-1bmnw, maybridge1_003980, 3-cyano-benzenesulfonamide, 3-cyanobenzenesulfonamide, benzenesulfonamide,3-cyano-9ci
Formula Weight 182.2
Percent Purity 98%
Chemical Name or Material 3-Cyanobenzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'