missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzoic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11470308
Click to view available options
Quantity:
5 g
25 g
100 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1877-72-1
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.133
MDL Number MFCD00002486
InChI Key GYLKKXHEIIFTJH-UHFFFAOYSA-N
Synonym m-cyanobenzoic acid, benzoic acid, 3-cyano, 3-carboxybenzonitrile, 3-cyano-benzoic acid, 3-cyanobenzoicacid, benzoic acid, m-cyano, unii-d2gc692d78, 3-cyano-benzoicacid, m-carboxybenzonitrile, 3-cyano benzoic acid
PubChem CID 15875
IUPAC Name 3-cyanobenzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C#N

Specifications

Melting Point 219°C to 224°C
Quantity 5 g
Beilstein 1862566
Formula Weight 147.13
Percent Purity ≥98%
Chemical Name or Material 3-Cyanobenzoic acid
compliance-icons
Product Identifier
  • 3-Cyanobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : m-Cyanobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'