Learn More
(3-Cyanopropyl)trichlorosilane, 98%, Thermo Scientific Chemicals
£56.38 - £155.78
Chemical Identifiers
CAS | 1071-27-8 |
---|---|
Molecular Formula | C4H6Cl3NSi |
Molecular Weight (g/mol) | 202.534 |
MDL Number | MFCD00013832 |
InChI Key | HMFFOEBLYHLRQN-UHFFFAOYSA-N |
Synonym | 3-cyanopropyltrichlorosilane, butanenitrile, 4-trichlorosilyl, 4-trichlorosilyl butyronitrile, 4-cyanobutyltrichlorosilane, butyronitrile, 4-trichlorosilyl, trichloro 3-cyanopropyl silane, trichlor-3-kyanpropylsilan, silane, trichloro 3-cyanopropyl, unii-05m8iu15r8, gamma-cyanopropyltrichlorosilane |
PubChem CID | 14063 |
IUPAC Name | 4-trichlorosilylbutanenitrile |
SMILES | C(CC#N)C[Si](Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499050
|
Thermo Scientific Alfa Aesar
A13787.14 |
25 g |
£56.38
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409060
|
Thermo Scientific Alfa Aesar
A13787.22 |
100 g |
£155.78
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1071-27-8 | |
202.534 | |
HMFFOEBLYHLRQN-UHFFFAOYSA-N | |
14063 | |
C(CC#N)C[Si](Cl)(Cl)Cl |
C4H6Cl3NSi | |
MFCD00013832 | |
3-cyanopropyltrichlorosilane, butanenitrile, 4-trichlorosilyl, 4-trichlorosilyl butyronitrile, 4-cyanobutyltrichlorosilane, butyronitrile, 4-trichlorosilyl, trichloro 3-cyanopropyl silane, trichlor-3-kyanpropylsilan, silane, trichloro 3-cyanopropyl, unii-05m8iu15r8, gamma-cyanopropyltrichlorosilane | |
4-trichlorosilylbutanenitrile |
Specifications
237°C to 238°C | |
C4H6Cl3NSi | |
MFCD00013832 | |
906924 | |
3-cyanopropyltrichlorosilane, butanenitrile, 4-trichlorosilyl, 4-trichlorosilyl butyronitrile, 4-cyanobutyltrichlorosilane, butyronitrile, 4-trichlorosilyl, trichloro 3-cyanopropyl silane, trichlor-3-kyanpropylsilan, silane, trichloro 3-cyanopropyl, unii-05m8iu15r8, gamma-cyanopropyltrichlorosilane | |
C(CC#N)C[Si](Cl)(Cl)Cl | |
202.534 | |
202.54 | |
Pungent | |
Moisture sensitive | |
(3-Cyanopropyl)trichlorosilane |
1071-27-8 | |
Cl3Si(CH2)3CN | |
UN2987 | |
92°C (198°F) | |
HMFFOEBLYHLRQN-UHFFFAOYSA-N | |
4-trichlorosilylbutanenitrile | |
14063 | |
98% | |
1.463 | |
1.28 |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 213-990-6
missing translation for 'rtecsNumber' : VV2775000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only