missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyanopropyltrichlorosilane, 97%, ACROS Organics™
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Specifications
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Color | Colorless |
Density | 1.3000g/mL |
Boiling Point | 237.0°C to 238.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Linear Formula | CN(CH2)3Si(Cl)3 |
Refractive Index | 1.4620 to 1.4640 (20°C, 589nm) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'