missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexene-1-carbonyl chloride, 96%, Thermo Scientific™

Product Code. 11365379
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Cyclohexene-1-carbonyl chloride is used as an active pharmaceutical ingredients.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 932-67-2
Molecular Formula C7H9ClO
Molecular Weight (g/mol) 144.60
MDL Number MFCD00013780
InChI Key CXWMHIRIXVNVQN-UHFFFAOYNA-N
Synonym 3-cyclohexene-1-carbonyl chloride, 3-cyclohexenecarbonyl chloride, cyclohex-3-enecarbonyl chloride, 3-cyclohexene-1-carbonylchloride, acmc-20an5a, 3-cyclohexenecarbonylchloride, 3-cyclohexene carbonyl chloride, 1-cyclohex-3-enecarbonyl chloride, 3-cyclohexene-1-carbonyl chloride #, 3-cyclohexenecarboxylic acid chloride
PubChem CID 523058
IUPAC Name cyclohex-3-ene-1-carbonyl chloride
SMILES ClC(=O)C1CCC=CC1

Specifications

Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 144.6
Percent Purity 96%
Chemical Name or Material 3-Cyclohexene-1-carbonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'