missing translation for 'onlineSavingsMsg'
Learn More

3-(Cyclopentylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific™

£33.00 - £109.00

Chemical Identifiers

CAS 850567-24-7
Molecular Formula C12H16BNO3
Molecular Weight (g/mol) 233.074
MDL Number MFCD04115695
InChI Key KIOBFDGZJVJNBP-UHFFFAOYSA-N
Synonym 3-cyclopentylaminocarbonyl phenylboronic acid, 3-cyclopentylcarbamoyl phenyl boronic acid, 3-cyclopentylcarbamoyl phenylboronic acid, 3-cyclopentylaminocarbonyl benzeneboronic acid, pubchem16391, acmc-209q0d, 3-cyclopentylcarbamoyl benzeneboronic acid, n-cyclopentyl-3-boronobenzamide
PubChem CID 3775808
IUPAC Name [3-(cyclopentylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)C(=O)NC2CCCC2)(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15440406
View Documents
Thermo Scientific Alfa Aesar
H53125.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15479887
View Documents
Thermo Scientific Alfa Aesar
H53125.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

850567-24-7
233.074
KIOBFDGZJVJNBP-UHFFFAOYSA-N
3775808
B(C1=CC(=CC=C1)C(=O)NC2CCCC2)(O)O
C12H16BNO3
MFCD04115695
3-cyclopentylaminocarbonyl phenylboronic acid, 3-cyclopentylcarbamoyl phenyl boronic acid, 3-cyclopentylcarbamoyl phenylboronic acid, 3-cyclopentylaminocarbonyl benzeneboronic acid, pubchem16391, acmc-209q0d, 3-cyclopentylcarbamoyl benzeneboronic acid, n-cyclopentyl-3-boronobenzamide
[3-(cyclopentylcarbamoyl)phenyl]boronic acid

Specifications

850567-24-7
MFCD04115695
KIOBFDGZJVJNBP-UHFFFAOYSA-N
[3-(cyclopentylcarbamoyl)phenyl]boronic acid
3775808
98%
C12H16BNO3
3-cyclopentylaminocarbonyl phenylboronic acid, 3-cyclopentylcarbamoyl phenyl boronic acid, 3-cyclopentylcarbamoyl phenylboronic acid, 3-cyclopentylaminocarbonyl benzeneboronic acid, pubchem16391, acmc-209q0d, 3-cyclopentylcarbamoyl benzeneboronic acid, n-cyclopentyl-3-boronobenzamide
B(C1=CC(=CC=C1)C(=O)NC2CCCC2)(O)O
233.074
233.07
3-(Cyclopentylcarbamoyl)benzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.