missing translation for 'onlineSavingsMsg'
Learn More

3-(Cyclopentyloxy)-4-methoxybenzoic acid, 97%, Thermo Scientific™

Product Code. 10314442
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 144036-17-9
Molecular Formula C13H16O4
Molecular Weight (g/mol) 236.27
MDL Number MFCD00219797
InChI Key RVADCQWIQKYXBJ-UHFFFAOYSA-N
Synonym 3-cyclopentyloxy-4-methoxybenzoic acid, benzoic acid,3-cyclopentyloxy-4-methoxy, 3-cyclopentyloxy-4-methoxy benzoic acid, maybridge1_005004, acmc-20e1kk, 3-cyclopentyloxy-4-methoxy-benzoic acid, 3-cyclopentoxy-4-methoxy-benzoic acid, 3-cyclopentyloxy-4-methoxybezoic acid, 3-cyclopentyl-oxy-4-methoxybenzoic acid
PubChem CID 2779323
IUPAC Name 3-cyclopentyloxy-4-methoxybenzoic acid
SMILES COC1=C(OC2CCCC2)C=C(C=C1)C(O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 236.27
Percent Purity 97%
Chemical Name or Material 3-(Cyclopentyloxy)-4-methoxybenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'