missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopentylpropionic acid, 98%, Thermo Scientific Chemicals

Product Code. 10395930
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-77-2
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00001392
InChI Key ZRPLANDPDWYOMZ-UHFFFAOYSA-N
Synonym 3-cyclopentylpropionic acid, cyclopentanepropanoic acid, cyclopentanepropionic acid, 3-cylcopentylpropanoic acid, cyclopentylpropionic acid, propionic acid, 3-cyclopentyl, 3-cyclopentyl-propionic acid, cypionic acid, 3-cyclopentylpropionicacid, cipionate
PubChem CID 8818
ChEBI CHEBI:50899
IUPAC Name 3-cyclopentylpropanoic acid
SMILES OC(=O)CCC1CCCC1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 130.0°C to 132.0°C (12.0 mmHg)
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C5H9CH2CH2CO2H
Refractive Index 1.4560 to 1.4580
Quantity 25 g
Specific Gravity 0.99
Solubility Information Solubility in water: insoluble
Formula Weight 142.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Cyclopentylpropionic acid, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'