Learn More
3-Cyclopropyl-1-methyl-1H-pyrazol-5-amine, 97%, Thermo Scientific™
£8.10 - £112.00
Chemical Identifiers
CAS | 118430-74-3 |
---|---|
Molecular Formula | C7H11N3 |
Molecular Weight (g/mol) | 137.19 |
MDL Number | MFCD00067984 |
InChI Key | YWHWPIRLFHZSFS-UHFFFAOYSA-N |
PubChem CID | 2735311 |
SMILES | CN1N=C(C=C1N)C1CC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10466272
|
Thermo Scientific Maybridge
KM00278DA |
1 g | Amber Glass Bottle |
£8.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10553022
|
Thermo Scientific Maybridge
KM00278EA |
10 g | Amber Glass Bottle |
£57.50
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10727115
|
Thermo Scientific Maybridge
KM00278EB |
25 g | Amber Glass Bottle |
£112.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
118430-74-3 | |
137.19 | |
YWHWPIRLFHZSFS-UHFFFAOYSA-N | |
CN1N=C(C=C1N)C1CC1 |
C7H11N3 | |
MFCD00067984 | |
2735311 |
Specifications
118430-74-3 | |
97% | |
MFCD00067984 | |
YWHWPIRLFHZSFS-UHFFFAOYSA-N | |
5-cyclopropyl-2-methylpyrazol-3-amine | |
2735311 | |
97% |
97.0 | |
C7H11N3 | |
3-cyclopropyl-1-methyl-1h-pyrazol-5-amine, 1h-pyrazol-5-amine, 3-cyclopropyl-1-methyl, 5-amino-3-cyclopropyl-1-methylpyrazole, 5-amino-3-cyclopropyl-1-methyl-1h-pyrazole, 5-cyclopropyl-2-methyl-2h-pyrazol-3-ylamine, 1h-pyrazol-5-amine,3-cyclopropyl-1-methyl, 3-cyclopropyl-1-methylpyrazole-5-ylamine, acmc-1cbj7, 5-amino-3-cyclopropyl-1-methyl pyrazole, 5-cyclopropyl-2-methyl-pyrazole-3-amine | |
CN1N=C(C=C1N)C1CC1 | |
137.19 | |
137.18 | |
3-Cyclopropyl-1-methyl-1H-pyrazol-5-amine |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.