missing translation for 'onlineSavingsMsg'
Learn More
3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid, 98%, Thermo Scientific Chemicals Product Code.: 15488696

3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 15488696
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15488696

Brand: Thermo Scientific Alfa Aesar H64098.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162401-62-9
Molecular Formula C12H12F2O4
Molecular Weight (g/mol) 258.221
MDL Number MFCD04621687
InChI Key IGFDIFLMMLWKKY-UHFFFAOYSA-N
Synonym 3-cyclopropylmethoxy-4-difluoromethoxy benzoic acid, 3-cyclopropylmethoxy-4-difluoromethoxy-benzoic acid, benzoic acid, 3-cyclopropylmethoxy-4-difluoromethoxy, roflumilast intermediate, roflumilast intermediate b, 3-cyclopropylmethoxy-4-difluoromethoxybenzoicacid, 3-cyclopropylmethoxy-4-difluoromethoxy-benzoic acid, 3-cyclopropylmethoxy 4-difluoromethoxy benzoic acid
PubChem CID 2394006
IUPAC Name 3-(cyclopropylmethoxy)-4-(difluoromethoxy)benzoic acid
SMILES C1CC1COC2=C(C=CC(=C2)C(=O)O)OC(F)F

Specifications

Melting Point 118°C to 120°C
Quantity 5 g
Beilstein 11413145
Formula Weight 258.22
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP20J011 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberH64098.03,H64098.06
Lot Number5010J09N Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH64098.03,H64098.06,H64098.MD
Lot Number81302481 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH64098.03,H64098.06,H64098.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.