missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3'-Deoxythymidine

Product Code. 15417069
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
This item is not returnable. View return policy

Product Code. 15417069

Brand: Thermo Scientific Chemicals J65765.ME

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3416-05-5 | C10H14N2O4 | 226.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3416-05-5
Molecular Formula C10H14N2O4
Molecular Weight (g/mol) 226.23
MDL Number MFCD00010570
InChI Key XKKCQTLDIPIRQD-JGVFFNPUSA-N
Synonym 3'-deoxythymidine, 2',3'-dideoxythymidine, 1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidine-2,4 1h,3h-dione, dideoxythymidine, thymidine, 3'-deoxy, desoxythymidin, 1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-5-methyl-pyrimidine-2,4-dione, ddt & gm-csf
PubChem CID 65119
IUPAC Name 1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN([C@H]2CC[C@@H](CO)O2)C(=O)NC1=O

Specifications

Melting Point 155°C to 156°C
Color White to Yellow
Quantity 500 mg
Beilstein 21884
Formula Weight 226.23
Physical Form Powder
Chemical Name or Material 3'-Deoxythymidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.