missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylamino-2-methylpropyl chloride hydrochloride, 98%, Thermo Scientific™

£11.05 - £121.00

Chemical Identifiers

CAS 4261-67-0
Molecular Formula C6H15Cl2N
Molecular Weight (g/mol) 172.093
MDL Number MFCD00012514
InChI Key SOMIBONUMGNAEP-UHFFFAOYSA-N
Synonym 3-dimethylamino-2-methylpropyl chloride hydrochloride, 3-chloro-n,n,2-trimethylpropan-1-amine hydrochloride, 3-chloro-2-methylpropyl dimethyl ammonium chloride, 3-chloro-2-methylpropyl dimethylamine hydrochloride, acmc-20an51, 3-dimethylamino-2-methylpropyl chloride hcl, 3-chloro-n,n,2-trimethylpropylamine hydrochloride, 2-methyl-3-dimethylaminopropylchloride hydrochloride, 3-dimethylamino-2-methylpropylchloride hydrochloride, 1-dimethylamino-2-methyl-3-chloropropane hydrochloride
PubChem CID 3083841
IUPAC Name 3-chloro-N,N,2-trimethylpropan-1-amine;hydrochloride
SMILES CC(CN(C)C)CCl.Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11342366
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L06091.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11352366
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L06091.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11362366
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L06091.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4261-67-0
172.093
SOMIBONUMGNAEP-UHFFFAOYSA-N
3083841
CC(CN(C)C)CCl.Cl
C6H15Cl2N
MFCD00012514
3-dimethylamino-2-methylpropyl chloride hydrochloride, 3-chloro-n,n,2-trimethylpropan-1-amine hydrochloride, 3-chloro-2-methylpropyl dimethyl ammonium chloride, 3-chloro-2-methylpropyl dimethylamine hydrochloride, acmc-20an51, 3-dimethylamino-2-methylpropyl chloride hcl, 3-chloro-n,n,2-trimethylpropylamine hydrochloride, 2-methyl-3-dimethylaminopropylchloride hydrochloride, 3-dimethylamino-2-methylpropylchloride hydrochloride, 1-dimethylamino-2-methyl-3-chloropropane hydrochloride
3-chloro-N,N,2-trimethylpropan-1-amine;hydrochloride

Specifications

4261-67-0
C6H15Cl2N
4444785
3-dimethylamino-2-methylpropyl chloride hydrochloride, 3-chloro-n,n,2-trimethylpropan-1-amine hydrochloride, 3-chloro-2-methylpropyl dimethyl ammonium chloride, 3-chloro-2-methylpropyl dimethylamine hydrochloride, acmc-20an51, 3-dimethylamino-2-methylpropyl chloride hcl, 3-chloro-n,n,2-trimethylpropylamine hydrochloride, 2-methyl-3-dimethylaminopropylchloride hydrochloride, 3-dimethylamino-2-methylpropylchloride hydrochloride, 1-dimethylamino-2-methyl-3-chloropropane hydrochloride
CC(CN(C)C)CCl.Cl
172.093
172.1
3-Dimethylamino-2-methylpropyl chloride hydrochloride
167°C to 170°C
MFCD00012514
Hygroscopic
SOMIBONUMGNAEP-UHFFFAOYSA-N
3-chloro-N,N,2-trimethylpropan-1-amine;hydrochloride
3083841
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 224-237-6

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications