missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylamino)benzyl alcohol, 97%, Thermo Scientific Chemicals

Product Code. 10047659
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23501-93-1
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00017342
InChI Key YTUXZVSDDHCTBZ-UHFFFAOYSA-N
Synonym 3-dimethylamino phenyl methanol, 3-dimethylamino benzyl alcohol, 3-dimethylaminobenzyl alcohol, m-dimethylamino benzyl alcohol, benzenemethanol,3-dimethylamino, 3-dimethylamino phenyl methan-1-ol, benzyl alcohol, m-dimethylamino, benzenemethanol, 3-dimethylamino
PubChem CID 90133
IUPAC Name [3-(dimethylamino)phenyl]methanol
SMILES CN(C)C1=CC=CC(=C1)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 109.0°C to 110.0°C (0.1 Torr)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 151.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-(Dimethylamino)benzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 3-(Dimethylamino)benzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'