missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-2-cyclohexen-1-one, 99%, Thermo Scientific Chemicals

Product Code. 10509430
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5323-87-5
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00001580
InChI Key JWCFJPLIRVYENQ-UHFFFAOYSA-N
Synonym 3-ethoxy-2-cyclohexen-1-one, 3-ethoxycyclohex-2-enone, 2-cyclohexen-1-one, 3-ethoxy, 3-ethoxy-2-cyclohexenone, 3-ethoxy-2-cyclohexene-1-one, 3-ethoxycyclohex-2-ene-1-one, 1-ethoxycyclohexene-3-one, acmc-209l4y, 3-ethoxy-2-cyclohexen-one, 3-ethoxy-cyclohex-2-enone
PubChem CID 79216
IUPAC Name 3-ethoxycyclohex-2-en-1-one
SMILES CCOC1=CC(=O)CCC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Peach to White
Density 1.0400g/mL
Boiling Point 76°C to 78°C (1.0 mmHg)
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5OC6H7(=O)
Refractive Index 1.5030 to 1.505
Quantity 10 g
Specific Gravity 1.04
Solubility Information Solubility in water: slightly soluble
Formula Weight 140.18
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethoxy-2-cyclohexen-1-one
Show More Show Less
Product Identifier
  • 3-Ethoxy-2-cyclohexen-1-one

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'