Learn More
3-Ethoxy-2-nitropyridine, 98%, Thermo Scientific™
£22.70 - £339.00
Chemical Identifiers
CAS | 74037-50-6 |
---|---|
Molecular Formula | C7H8N2O3 |
Molecular Weight (g/mol) | 168.15 |
MDL Number | MFCD00006257 |
InChI Key | WBEVIQNYYGPESH-UHFFFAOYSA-N |
PubChem CID | 52866 |
IUPAC Name | 3-ethoxy-2-nitropyridine |
SMILES | CCOC1=CC=CN=C1[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11439090
|
Thermo Scientific Alfa Aesar
A14371.03 |
1 g | |||||||
11449090
|
Thermo Scientific Alfa Aesar
A14371.06 |
5 g | |||||||
11459090
|
Thermo Scientific Alfa Aesar
A14371.09 |
10 g | |||||||
11469090
|
Thermo Scientific Alfa Aesar
A14371.14 |
25 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
74037-50-6 | |
168.15 | |
WBEVIQNYYGPESH-UHFFFAOYSA-N | |
3-ethoxy-2-nitropyridine |
C7H8N2O3 | |
MFCD00006257 | |
52866 | |
CCOC1=CC=CN=C1[N+]([O-])=O |
Specifications
74037-50-6 | |
156°C to 158°C (15 mmHg) | |
C7H8N2O3 | |
141592 | |
WBEVIQNYYGPESH-UHFFFAOYSA-N | |
3-ethoxy-2-nitropyridine | |
52866 | |
98% |
28°C to 32°C | |
>100°C (212°F) | |
MFCD00006257 | |
pyridine, 3-ethoxy-2-nitro, 3-ethoxy-2-nitro-pyridine, 2-nitro-3-ethoxy pyridine, pubchem6624, acmc-209otd, 3-ethoxy-2-nitropyridin, 2-nitro-3-ethoxypyridine, pyridine,3-ethoxy-2-nitro, 4-21-00-00436 beilstein handbook reference | |
CCOC1=CC=CN=C1[N+]([O-])=O | |
168.15 | |
168.15 | |
3-Ethoxy-2-nitropyridine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H228-H302-H315-H319-H335-H500
missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: II
missing translation for 'einecsNumber' : 277-676-0
missing translation for 'rtecsNumber' : UT3001500
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures