missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-2-nitropyridine, 98%, Thermo Scientific™

Product Code. 11439090
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74037-50-6
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.15
MDL Number MFCD00006257
InChI Key WBEVIQNYYGPESH-UHFFFAOYSA-N
PubChem CID 52866
IUPAC Name 3-ethoxy-2-nitropyridine
SMILES CCOC1=CC=CN=C1[N+]([O-])=O

Specifications

Melting Point 28°C to 32°C
Boiling Point 156°C to 158°C (15 mmHg)
Flash Point >100°C (212°F)
Quantity 1 g
Beilstein 141592
Synonym pyridine, 3-ethoxy-2-nitro, 3-ethoxy-2-nitro-pyridine, 2-nitro-3-ethoxy pyridine, pubchem6624, acmc-209otd, 3-ethoxy-2-nitropyridin, 2-nitro-3-ethoxypyridine, pyridine,3-ethoxy-2-nitro, 4-21-00-00436 beilstein handbook reference
Formula Weight 168.15
Percent Purity 98%
Chemical Name or Material 3-Ethoxy-2-nitropyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'