missing translation for 'onlineSavingsMsg'
Learn More

3-(Ethoxycarbonyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

£63.90 - £208.00

Chemical Identifiers

CAS 4334-87-6
Molecular Formula C9H11BO4
Molecular Weight (g/mol) 193.99
MDL Number MFCD02179444
InChI Key REHVCPNQQBDOJJ-UHFFFAOYSA-N
Synonym 3-ethoxycarbonyl phenylboronic acid, 3-ethoxycarbonylphenyl boronic acid, 3-carbethoxyphenylboronic acid, m-ethoxycarbonylphenylboronic acid, 3-ethoxycarbonyl phenyl boronic acid, ethyl 3-dihydroxyboranyl benzoate, ethyl 3-boronobenzoate, 3-ethoxycarbonylbenzeneboronic acid, 3-ethoxycarbonyl benzeneboronic acid
PubChem CID 3249312
IUPAC Name (3-ethoxycarbonylphenyl)boronic acid
SMILES CCOC(=O)C1=CC=CC(=C1)B(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11342479
View Documents
Thermo Scientific Alfa Aesar
H26615.03
1 g
£63.90
1g
Please to purchase this item. Need a web account? Register with us today!
11352479
View Documents
Thermo Scientific Alfa Aesar
H26615.06
5 g
£208.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4334-87-6
193.99
REHVCPNQQBDOJJ-UHFFFAOYSA-N
3249312
CCOC(=O)C1=CC=CC(=C1)B(O)O
C9H11BO4
MFCD02179444
3-ethoxycarbonyl phenylboronic acid, 3-ethoxycarbonylphenyl boronic acid, 3-carbethoxyphenylboronic acid, m-ethoxycarbonylphenylboronic acid, 3-ethoxycarbonyl phenyl boronic acid, ethyl 3-dihydroxyboranyl benzoate, ethyl 3-boronobenzoate, 3-ethoxycarbonylbenzeneboronic acid, 3-ethoxycarbonyl benzeneboronic acid
(3-ethoxycarbonylphenyl)boronic acid

Specifications

4334-87-6
C9H11BO4
3-ethoxycarbonyl phenylboronic acid, 3-ethoxycarbonylphenyl boronic acid, 3-carbethoxyphenylboronic acid, m-ethoxycarbonylphenylboronic acid, 3-ethoxycarbonyl phenyl boronic acid, ethyl 3-dihydroxyboranyl benzoate, ethyl 3-boronobenzoate, 3-ethoxycarbonylbenzeneboronic acid, 3-ethoxycarbonyl benzeneboronic acid
CCOC(=O)C1=CC=CC(=C1)B(O)O
193.99
193.99
3-(Ethoxycarbonyl)benzeneboronic acid
135°C to 139°C
MFCD02179444
REHVCPNQQBDOJJ-UHFFFAOYSA-N
(3-ethoxycarbonylphenyl)boronic acid
3249312
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only