missing translation for 'onlineSavingsMsg'
Learn More

3-(Ethylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15469437
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850567-21-4
Molecular Formula C9H12BNO3
Molecular Weight (g/mol) 193.01
MDL Number MFCD04115689
InChI Key WMMRKCBSJZVRRG-UHFFFAOYSA-N
Synonym 3-n-ethylaminocarbonyl phenylboronic acid, 3-n-ethylaminocarbonyl benzeneboronic acid, 3-ethylcarbamoyl phenylboronic acid, 3-ethylcarbamoyl benzeneboronic acid, 3-ethylcarbamoyl phenyl boronic acid, 3-n-ethylaminocarboyl phenylboronic acid, boronic acid,b-3-ethylamino carbonyl phenyl, acmc-209q0a, 3-ethylcarbamoyl benzeneboronic acid
PubChem CID 3607123
IUPAC Name [3-(ethylcarbamoyl)phenyl]boronic acid
SMILES CCNC(=O)C1=CC=CC(=C1)B(O)O

Specifications

Melting Point 106°C to 116°C
Quantity 1 g
Formula Weight 193.01
Percent Purity 97%
Chemical Name or Material 3-(Ethylcarbamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'