missing translation for 'onlineSavingsMsg'
Learn More
3-Ethylcyclopentanone, 99%, Thermo Scientific Chemicals Product Code.: 11404387

3-Ethylcyclopentanone, 99%, Thermo Scientific Chemicals

Product Code. 11404387
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11404387

Brand: Thermo Scientific Alfa Aesar B20432.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Ethylcyclopentanone, is used as a chemical reagent, pharmaceutical reagent and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethylcyclopentanone, is used as a chemical reagent, pharmaceutical reagent and pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10264-55-8
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00045445
InChI Key XERALSLWOPMNRJ-UHFFFAOYSA-N
PubChem CID 139121
IUPAC Name 3-ethylcyclopentan-1-one
SMILES CCC1CCC(=O)C1

Specifications

Boiling Point 171°C
Flash Point 31°C (88°F)
Quantity 1 g
UN Number UN1224
Synonym 3-ethylcyclopentanone, cyclopentanone, 3-ethyl, 3-ethyl cyclopentanone, cyclopentanone,3-ethyl, acmc-20an48, 3-ethyl-cyclopentan-1-one
Solubility Information Soluble in water.
Formula Weight 112.17
Percent Purity 99%
Chemical Name or Material 3-Ethylcyclopentanone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Ethylcyclopentanone, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.