missing translation for 'onlineSavingsMsg'
Learn More

3-Ethynylthiophene, 96%, Thermo Scientific Chemicals

Product Code. 15492288
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67237-53-0
Molecular Formula C6H4S
Molecular Weight (g/mol) 108.158
MDL Number MFCD04039973
InChI Key MJHLPKWONJUCFK-UHFFFAOYSA-N
Synonym 3-ethynyl thiophene, thiophene, 3-ethynyl, 3-thienylacetylene, thiophen-3-yl acetylene, 3-ethynyl-thiophene, 3-ethynyl thiophene;, pubchem16122, thien-3-yl acetylene, 3-ethynylthiophene, ksc352m9b
PubChem CID 3548422
IUPAC Name 3-ethynylthiophene
SMILES C#CC1=CSC=C1

Specifications

Density 1.098
Boiling Point 152°C to 153°C
Flash Point 46°C (115°F)
Refractive Index 1.58
Quantity 5 g
UN Number UN1993
Solubility Information Slightly soluble in water.
Formula Weight 108.16
Percent Purity 96%
Chemical Name or Material 3-Ethynylthiophene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'