Learn More
3-Ethynylthiophene, 96%, Thermo Scientific Chemicals
£20.60 - £176.00
Chemical Identifiers
CAS | 67237-53-0 |
---|---|
Molecular Formula | C6H4S |
Molecular Weight (g/mol) | 108.158 |
MDL Number | MFCD04039973 |
InChI Key | MJHLPKWONJUCFK-UHFFFAOYSA-N |
Synonym | 3-ethynyl thiophene, thiophene, 3-ethynyl, 3-thienylacetylene, thiophen-3-yl acetylene, 3-ethynyl-thiophene, 3-ethynyl thiophene;, pubchem16122, thien-3-yl acetylene, 3-ethynylthiophene, ksc352m9b |
PubChem CID | 3548422 |
IUPAC Name | 3-ethynylthiophene |
SMILES | C#CC1=CSC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15402298
|
Thermo Scientific Alfa Aesar
H55898.MD |
250 mg |
£20.60
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15482288
|
Thermo Scientific Alfa Aesar
H55898.03 |
1 g |
£55.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15492288
|
Thermo Scientific Alfa Aesar
H55898.06 |
5 g |
£176.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water.
Notes
Store at room temperature. Avoid strong oxidizing agents.
Chemical Identifiers
67237-53-0 | |
108.158 | |
MJHLPKWONJUCFK-UHFFFAOYSA-N | |
3548422 | |
C#CC1=CSC=C1 |
C6H4S | |
MFCD04039973 | |
3-ethynyl thiophene, thiophene, 3-ethynyl, 3-thienylacetylene, thiophen-3-yl acetylene, 3-ethynyl-thiophene, 3-ethynyl thiophene;, pubchem16122, thien-3-yl acetylene, 3-ethynylthiophene, ksc352m9b | |
3-ethynylthiophene |
Specifications
67237-53-0 | |
152°C to 153°C | |
C6H4S | |
MFCD04039973 | |
3-ethynyl thiophene, thiophene, 3-ethynyl, 3-thienylacetylene, thiophen-3-yl acetylene, 3-ethynyl-thiophene, 3-ethynyl thiophene;, pubchem16122, thien-3-yl acetylene, 3-ethynylthiophene, ksc352m9b | |
MJHLPKWONJUCFK-UHFFFAOYSA-N | |
3-ethynylthiophene | |
3548422 | |
96% |
1.098 | |
46°C (115°F) | |
1.58 | |
UN1993 | |
Slightly soluble in water. | |
C#CC1=CSC=C1 | |
108.158 | |
108.16 | |
3-Ethynylthiophene |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H318-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only