Learn More
Bochem™ 3-Finger Laboral Retort Clamp
Coated with soft PVC, the three fingers of this independently adjustable retort clamp are well suited for holding delicate objects. Bochem™ 3-Finger Laboral Retort Clamp has a base construction of Laboral aluminum alloy.
Brand:  Bochem™ B08256108
Product Code. 10228980
Description
Description
  • Laboral aluminum alloy construction.
  • PVC-coated fingers.
  • Span width is independently adjustable with 2 regulating screws.
Specifications
Specifications
1 Ea.
Laboral, PVC
12 mm
140 mm
3-Prong
Stand Clamp
12 to 100 mm
140 mm
12 to 100 mm
12 to 100 mm