missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-formylbenzeneboronic acid, 95%, Thermo Scientific Chemicals

Product Code. 15482319
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Undergoes unprecedented tautomeric rearrangement in solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Undergoes unprecedented tautomeric rearrangement in solution.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871126-15-7
Molecular Formula C7H6BFO3
Molecular Weight (g/mol) 167.93
MDL Number MFCD10697421
InChI Key GVHWLCYABLWGIR-UHFFFAOYSA-N
Synonym 3-fluoro-2-formylphenyl boronic acid, 3-fluoro-2-formylbenzeneboronic acid, boronic acid, 3-fluoro-2-formylphenyl, acmc-209qf3, 2-formyl-3-fluorophenylboranic acid, 3-fluoro-2-formyl-phenyl boronic acid, 3-fluoro-2-formylphenylboronic acid, boronic acid, b-3-fluoro-2-formylphenyl, contains varying amounts of anhydride
PubChem CID 53412038
IUPAC Name (3-fluoro-2-formylphenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)F)C=O)(O)O

Specifications

Melting Point 123°C to 127°C
Quantity 25 g
UN Number UN3261
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 167.93
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Fluoro-2-formylbenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'