missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-methylbenzoic acid, 98%, Thermo Scientific™

Product Code. 15454547
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 699-90-1
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00075246
InChI Key XMKZAIHFVHJGPV-UHFFFAOYSA-N
Synonym 3-fluoro-2-methyl-benzoic acid, 2-methyl-3-fluoro benzoic acid, 3-fluoro-2-methylbenzoicacid, benzoic acid, 3-fluoro-2-methyl, 6-fluortoluylsaure, pubchem1329, 2-carboxy-6-fluorotoluene, acmc-209oc1, ksc352q5t, 2-methyl-3-fluorobenzoic acid
PubChem CID 246682
IUPAC Name 3-fluoro-2-methylbenzoic acid
SMILES CC1=C(C=CC=C1F)C(=O)O

Specifications

Quantity 1 g
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 3-Fluoro-2-methylbenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'