missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-hydroxy-5-nitrobenzaldehyde, 95%, Thermo Scientific™

Product Code. 15438377
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 711022-47-8
Molecular Formula C7H4FNO4
Molecular Weight (g/mol) 185.11
MDL Number MFCD11846408
InChI Key CZOMOYVUYOVNHX-UHFFFAOYSA-N
PubChem CID 18452164
IUPAC Name 3-fluoro-4-hydroxy-5-nitrobenzaldehyde
SMILES OC1=C(C=C(C=O)C=C1F)[N+]([O-])=O

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 185.11
Percent Purity 95%
Chemical Name or Material 3-Fluoro-4-hydroxy-5-nitrobenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'