Learn More
3-Fluoro-4-methoxyaniline, 99%, Thermo Scientific Chemicals
£36.60 - £123.00
Chemical Identifiers
CAS | 366-99-4 |
---|---|
Molecular Formula | C7H8FNO |
Molecular Weight (g/mol) | 141.15 |
MDL Number | MFCD00075040 |
InChI Key | LJWAPDSCYTZUJU-UHFFFAOYSA-N |
Synonym | 3-fluoro-4-anisidine, 4-amino-2-fluoroanisole, 3-fluoro-p-anisidine, 3-fluoro-4-methoxy-phenylamine, benzenamine, 3-fluoro-4-methoxy, 3-fluoro-4-methoxy-aniline, 3-fluoro-4-methoxyphenylamine, 2-fluoro-4-aminoanisole, 3-fluoro-4-methoxy-anilin, 3-fluoro-4-methoxy-benzenamine |
PubChem CID | 581110 |
IUPAC Name | 3-fluoro-4-methoxyaniline |
SMILES | COC1=C(C=C(C=C1)N)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10188942
|
Thermo Scientific Acros
375880010 |
1 g | Glass bottle |
£36.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10322433
|
Thermo Scientific
375880050 |
5 g | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
366-99-4 | |
141.15 | |
LJWAPDSCYTZUJU-UHFFFAOYSA-N | |
581110 | |
COC1=C(C=C(C=C1)N)F |
C7H8FNO | |
MFCD00075040 | |
3-fluoro-4-anisidine, 4-amino-2-fluoroanisole, 3-fluoro-p-anisidine, 3-fluoro-4-methoxy-phenylamine, benzenamine, 3-fluoro-4-methoxy, 3-fluoro-4-methoxy-aniline, 3-fluoro-4-methoxyphenylamine, 2-fluoro-4-aminoanisole, 3-fluoro-4-methoxy-anilin, 3-fluoro-4-methoxy-benzenamine | |
3-fluoro-4-methoxyaniline |
Specifications
366-99-4 | |
100.0 | |
Beige to Brown-Yellow | |
Authentic | |
C7H8FNO | |
MFCD00075040 | |
LJWAPDSCYTZUJU-UHFFFAOYSA-N | |
3-fluoro-4-methoxyaniline | |
581110 | |
99% | |
3-Fluoro-4-methoxyaniline |
98.5 | |
81°C to 84°C | |
135°C (18.0 mmHg) | |
98.5% min. (GC) | |
FC6H3(NH2)OCH3 | |
3-fluoro-4-anisidine, 4-amino-2-fluoroanisole, 3-fluoro-p-anisidine, 3-fluoro-4-methoxy-phenylamine, benzenamine, 3-fluoro-4-methoxy, 3-fluoro-4-methoxy-aniline, 3-fluoro-4-methoxyphenylamine, 2-fluoro-4-aminoanisole, 3-fluoro-4-methoxy-anilin, 3-fluoro-4-methoxy-benzenamine | |
COC1=C(C=C(C=C1)N)F | |
141.15 | |
141.15 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact
GHS Signal Word: Warning
- 3-Fluoro-4-methoxyaniline
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only