missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methoxyphenylboronic acid, 97%, Thermo Scientific™

Product Code. 10647400
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 149507-26-6
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.95
InChI Key IILGLPAJXQMKGQ-UHFFFAOYSA-N
Synonym 3-fluoro-4-methoxybenzeneboronic acid, 3-fluoro-4-methoxyphenyl boronic acid, 3-fluoro-4-methoxyphenyl boranediol, 3-fluoro-4-methoxyphenylboronicacid, 4-methoxy-3-fluorophenylboronic acid, boronic acid, 3-fluoro-4-methoxyphenyl, 3-fluoro-4-methoxybenzeneboronicacid, pubchem1850, 3-fluoro-4-methoxy-benzeneboronic acid
PubChem CID 2782194
IUPAC Name (3-fluoro-4-methoxyphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)OC)F)(O)O

Especificaciones

Melting Point 211.0°C to 213.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol and thf
Formula Weight 169.95
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-methoxyphenylboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'