missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Fluoro-4-methylaniline, 98+%

Product Code. 11481940
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 452-77-7
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.146
MDL Number MFCD00007762
InChI Key MGRHBBRSAFPBIN-UHFFFAOYSA-N
Synonym 4-amino-2-fluorotoluene, 3-fluoro-p-toluidine, 2-fluoro-4-aminotoluene, 2-fluoro-4-amino-toluene, benzenamine, 3-fluoro-4-methyl, p-toluidine, 3-fluoro, 3-fluoro-4-methylphenylamine, 3-fluoro-4-methyl aniline, 3-fluoro-4-methyl-aniline, benzenamine, 3-fluoro-4-methyl-9ci
PubChem CID 9957
IUPAC Name 3-fluoro-4-methylaniline
SMILES CC1=C(C=C(C=C1)N)F

Specifications

Melting Point 30°C to 35°C
Density 1.093
Boiling Point 93°C (12 mmHg)
Flash Point 88°C (190°F)
Odor Characteristic
Refractive Index 1.539
Quantity 5 g
UN Number UN1759
Beilstein 2715987
Formula Weight 125.15
Percent Purity ≥98%
Chemical Name or Material 3-Fluoro-4-methylaniline
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'