missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(methylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15450556
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 849833-86-9
Molecular Formula C8H9BFNO3
Molecular Weight (g/mol) 196.972
MDL Number MFCD08436026
InChI Key IVSXSBGNMOZTJR-UHFFFAOYSA-N
Synonym 3-fluoro-4-methylcarbamoyl phenyl boronic acid, n-methyl 4-borono-2-fluorobenzamide, 3-fluoro-4-methylcarbamoyl phenylboronic acid, 3-fluoro-4-methylcarbamoyl benzeneboronic acid, acmc-209pz8, synquest 6660-3-t7, n-methyl-4-borono-2-fluorobenzamide, 3-fluoro-4-methylcarbamoyl phenyl boronicacid, 3-fluoranyl-4-methylcarbamoyl phenyl boronic acid
PubChem CID 44717467
IUPAC Name [3-fluoro-4-(methylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC(=C(C=C1)C(=O)NC)F)(O)O

Specifications

Melting Point 173°C to 175°C
Quantity 250 mg
Formula Weight 196.97
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-(methylcarbamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'