missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-nitrobenzyl bromide, 97%, Thermo Scientific Chemicals

Product Code. 11366599
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131858-37-2
Molecular Formula C7H5BrFNO2
Molecular Weight (g/mol) 234.02
MDL Number MFCD03094237
InChI Key ZOZJSWIXPIVMRU-UHFFFAOYSA-N
Synonym 3-fluoro-4-nitrobenzyl bromide, 4-bromomethyl-2-fluoro-1-nitrobenzene, 4-bromomethyl-2-fluoronitrobenzene, benzene, 4-bromomethyl-2-fluoro-1-nitro, intermediates-zcf02012, acmc-1c6h7, 4-bromomethyl-2-fluoro-1-nitro-benzene, benzene,4-bromomethyl-2-fluoro-1-nitro, 4-bromomethyl-2-fluoranyl-1-nitro-benzene
PubChem CID 2774661
IUPAC Name 4-(bromomethyl)-2-fluoro-1-nitrobenzene
SMILES [O-][N+](=O)C1=C(F)C=C(CBr)C=C1

Specifications

Melting Point 46°C to 48°C
Quantity 5 g
UN Number UN3261
Formula Weight 234.02
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-nitrobenzyl bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'