missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-5-methylbenzoic acid, 97%, Thermo Scientific™
£56.90 - £187.00
Chemical Identifiers
CAS | 518070-19-4 |
---|---|
Molecular Formula | C8H7FO2 |
Molecular Weight (g/mol) | 154.14 |
MDL Number | MFCD03094313 |
InChI Key | XPUFVYIUPYNLPD-UHFFFAOYSA-N |
Synonym | 3-fluoro-5-methylbenzoicacid, 3-fluoro-5-methyl-benzoic acid, benzoic acid, 3-fluoro-5-methyl, pubchem1396, 3-carboxy-5-fluorotoluene, acmc-1b05f, ksc498a8l, 3-fluoro-5-methybenzoic acid, rarechem al bo 2238, 3-fluoro-5-methyl benzoic acid |
PubChem CID | 2778447 |
IUPAC Name | 3-fluoro-5-methylbenzoic acid |
SMILES | CC1=CC(=CC(=C1)C(=O)O)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIntermediate in organic syntheses.
Solubility
Soluble in water (25 deg C)
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
518070-19-4 | |
154.14 | |
XPUFVYIUPYNLPD-UHFFFAOYSA-N | |
2778447 | |
CC1=CC(=CC(=C1)C(=O)O)F |
C8H7FO2 | |
MFCD03094313 | |
3-fluoro-5-methylbenzoicacid, 3-fluoro-5-methyl-benzoic acid, benzoic acid, 3-fluoro-5-methyl, pubchem1396, 3-carboxy-5-fluorotoluene, acmc-1b05f, ksc498a8l, 3-fluoro-5-methybenzoic acid, rarechem al bo 2238, 3-fluoro-5-methyl benzoic acid | |
3-fluoro-5-methylbenzoic acid |
Specifications
518070-19-4 | |
C8H7FO2 | |
3-fluoro-5-methylbenzoicacid, 3-fluoro-5-methyl-benzoic acid, benzoic acid, 3-fluoro-5-methyl, pubchem1396, 3-carboxy-5-fluorotoluene, acmc-1b05f, ksc498a8l, 3-fluoro-5-methybenzoic acid, rarechem al bo 2238, 3-fluoro-5-methyl benzoic acid | |
XPUFVYIUPYNLPD-UHFFFAOYSA-N | |
3-fluoro-5-methylbenzoic acid | |
2778447 | |
97% |
141°C to 143°C | |
MFCD03094313 | |
Soluble in water (25 deg C) | |
CC1=CC(=CC(=C1)C(=O)O)F | |
154.14 | |
154.14 | |
3-Fluoro-5-methylbenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures