missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-methylbenzonitrile, 97%, Thermo Scientific™

Product Code. 11431410
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216976-30-6
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.141
MDL Number MFCD03094312
InChI Key GKUQCZSZQBEHCP-UHFFFAOYSA-N
Synonym 3-cyano-5-fluorotoluene, 3-fluoro-5-methyl-benzonitrile, 3-fluoro-5-methyl benzonitrile, 5-fluoro-3-methylbenzenecarbonitrile, 5-fluoro-3-methylbenzonitrile, pubchem8336, acmc-20a0ds, ksc497i6f, benzonitrile, 3-fluoro-5-methyl, benzonitrile, 3-fluoro-5-methyl-9ci
PubChem CID 2778446
IUPAC Name 3-fluoro-5-methylbenzonitrile
SMILES CC1=CC(=CC(=C1)C#N)F

Specifications

Melting Point 53°C to 56°C
Quantity 5 g
UN Number UN3439
Formula Weight 135.14
Percent Purity 97%
Chemical Name or Material 3-Fluoro-5-methylbenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'