Learn More
3-Fluoro-5-methylbenzonitrile, 97%, Thermo Scientific™
£50.10 - £151.00
Chemical Identifiers
CAS | 216976-30-6 |
---|---|
Molecular Formula | C8H6FN |
Molecular Weight (g/mol) | 135.141 |
MDL Number | MFCD03094312 |
InChI Key | GKUQCZSZQBEHCP-UHFFFAOYSA-N |
Synonym | 3-cyano-5-fluorotoluene, 3-fluoro-5-methyl-benzonitrile, 3-fluoro-5-methyl benzonitrile, 5-fluoro-3-methylbenzenecarbonitrile, 5-fluoro-3-methylbenzonitrile, pubchem8336, acmc-20a0ds, ksc497i6f, benzonitrile, 3-fluoro-5-methyl, benzonitrile, 3-fluoro-5-methyl-9ci |
PubChem CID | 2778446 |
IUPAC Name | 3-fluoro-5-methylbenzonitrile |
SMILES | CC1=CC(=CC(=C1)C#N)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
216976-30-6 | |
135.141 | |
GKUQCZSZQBEHCP-UHFFFAOYSA-N | |
2778446 | |
CC1=CC(=CC(=C1)C#N)F |
C8H6FN | |
MFCD03094312 | |
3-cyano-5-fluorotoluene, 3-fluoro-5-methyl-benzonitrile, 3-fluoro-5-methyl benzonitrile, 5-fluoro-3-methylbenzenecarbonitrile, 5-fluoro-3-methylbenzonitrile, pubchem8336, acmc-20a0ds, ksc497i6f, benzonitrile, 3-fluoro-5-methyl, benzonitrile, 3-fluoro-5-methyl-9ci | |
3-fluoro-5-methylbenzonitrile |
Specifications
216976-30-6 | |
C8H6FN | |
UN3439 | |
GKUQCZSZQBEHCP-UHFFFAOYSA-N | |
3-fluoro-5-methylbenzonitrile | |
2778446 | |
97% |
53°C to 56°C | |
MFCD03094312 | |
3-cyano-5-fluorotoluene, 3-fluoro-5-methyl-benzonitrile, 3-fluoro-5-methyl benzonitrile, 5-fluoro-3-methylbenzenecarbonitrile, 5-fluoro-3-methylbenzonitrile, pubchem8336, acmc-20a0ds, ksc497i6f, benzonitrile, 3-fluoro-5-methyl, benzonitrile, 3-fluoro-5-methyl-9ci | |
CC1=CC(=CC(=C1)C#N)F | |
135.141 | |
135.14 | |
3-Fluoro-5-methylbenzonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures