missing translation for 'onlineSavingsMsg'
Learn More

3'-Fluoro-5'-(trifluoromethoxy)acetophenone, 97%, Thermo Scientific™

Product Code. 15418835
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1352999-53-1
Molecular Formula C9H6F4O2
Molecular Weight (g/mol) 222.139
MDL Number MFCD18783230
InChI Key VOBVUHOUGRWDOZ-UHFFFAOYSA-N
Synonym 3'-fluoro-5'-trifluoromethoxy acetophenone, 1-3-fluoro-5-trifluoromethoxy phenyl ethanone
PubChem CID 66523561
IUPAC Name 1-[3-fluoro-5-(trifluoromethoxy)phenyl]ethanone
SMILES CC(=O)C1=CC(=CC(=C1)F)OC(F)(F)F

Specifications

Quantity 250 mg
Formula Weight 222.14
Percent Purity 97%
Chemical Name or Material 3'-Fluoro-5'-(trifluoromethoxy)acetophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'