missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-DL-tyrosine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
500 mg
Description
3-Fluoro-DL-tyrosine is used for tyrosine in the biosynthesis of proteins such as β-galactosidases (Escherichia coli), bacteriorhodopsin and intestinal microvillar enzymes, aminopeptidase N, to study the effect of halogenated tryosines on the proteins properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Fluoro-DL-tyrosine is used for tyrosine in the biosynthesis of proteins such as β-galactosidases (Escherichia coli), bacteriorhodopsin and intestinal microvillar enzymes, aminopeptidase N, to study the effect of halogenated tryosines on the proteins properties.
Solubility
Soluble in water (28 g/L).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
3-Fluoro-DL-tyrosine is used for tyrosine in the biosynthesis of proteins such as β-galactosidases (Escherichia coli), bacteriorhodopsin and intestinal microvillar enzymes, aminopeptidase N, to study the effect of halogenated tryosines on the proteins properties.
Solubility
Soluble in water (28 g/L).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∽280°C (decomposition) |
Beilstein | 3204804 |
Solubility Information | Soluble in water (28g/L). |
Formula Weight | 199.18 |
Percent Purity | 98% |
Quantity | 100 mg |
Chemical Name or Material | 3-Fluoro-DL-tyrosine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'